Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Rabbit anti-Sodium Pump Subunit alpha-2, Polyclonal,

Catalog No. AB9094IMI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AB9094IMI 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AB9094IMI Supplier MilliporeSigma™ Supplier No. AB9094I
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Specifically detects Sodium Pump Subunit alpha-2 in Human, Mouse, Rat samples, and it is validated for Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blotting

Sodium/potassium-transporting ATPase subunit alpha-2 (EC 3.6.3.9; UniProt P06686; also known as Na+/K+ -ATPase alpha 2 subunit, Na+/K+ ATPase alpha(+) subunit, Sodium pump subunit alpha-2) is encoded by the Atp1a2 (also known as RATATPA2) gene (Gene ID 24212) in rat. Mammalian Na+/K+ ATPase is a heteromultimeric complex composed of alpha, beta, and gamma subunits. The alpha subunit catalyzes the hydrolysis of ATP coupled with the exchange of Na+ and K+ across the plasma membrane. The alpha subunit contains ten transmembrane domains with both its N- and C-termini in the cytoplasm. More than 50 mutations have been identified affecting the human ATP1A2 and ATP1A3 genes that are known to cause rapid-onset Dystonia Parkinsonism, familial hemiplegic migraine, alternating hemiplegia of childhood, and variants of familial hemiplegic migraine with neurological complications including seizures and various mood disorders.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Sodium Pump Subunit alpha-2
Applications Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Classification Polyclonal
Formulation Purified rabbit polyclonal antibody in buffer containing 0.1M Tris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide.
Gene Accession No. P06686
Gene Symbols Atp1a2
Host Species Rabbit
Immunogen KLH-conjugated linear peptide corresponding to the second cytoplasmic domain of rat Sodium Pump Subunit alpha-2.
Purification Method Affinity Purified
Quantity 100 μL
Regulatory Status RUO
Research Discipline Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) NP_036637
Target Species Human, Mouse, Rat
Content And Storage Stable for one year at 2°C to 8°C from date of receipt.
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.