Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TADA2A, Rabbit, Polyclonal Antibody, Abnova™

Catalog No. 89119001 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-119-001 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-119-001 Supplier Abnova Corporation Supplier No. PAB17345
Only null left
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against synthetic peptide of TADA2A.

Many DNA-binding transcriptional activator proteins enhance the initiation rate of RNA polymerase II-mediated gene transcription by interacting functionally with the general transcription machinery bound at the basal promoter. Adaptor proteins are usually required for this activation, possibly to acetylate and destabilize nucleosomes, thereby relieving chromatin constraints at the promoter. The protein encoded by this gene is a transcriptional activator adaptor and has been found to be part of the PCAF histone acetylase complex. Two transcript variants encoding different isoforms have been identified for this gene. [provided by RefSeq

Specifications

Antigen TADA2A
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against synthetic peptide of TADA2A.
Dilution Western Blot (1:500-1:1000) ELISA (1:10000) The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.4 (150mM NaCl, 0.02% sodium azide, 50% glycerol)
Gene TADA2L
Gene Alias ADA2/ADA2A/FLJ12705/KL04P/hADA2
Gene Symbols TADA2L
Host Species Rabbit
Immunogen A synthetic peptide corresponding to internal of human TADA2A.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6871
Test Specificity This antibody detects endogenous levels of total TADA2A protein.
Target Species Human, Mouse, Rat
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.