Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TLR4 Rabbit anti-Human, Polyclonal Antibody, Abnova™

Catalog No. 89134082 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-134-082 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-134-082 Supplier Abnova Corporation Supplier No. PAB8997
Only null left
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against synthetic peptide of TLR4.

The protein encoded by this gene is a member of the Toll-like receptor (TLR) family which plays a fundamental role in pathogen recognition and activation of innate immunity. TLRs are highly conserved from Drosophila to humans and share structural and functional similarities. They recognize pathogen-associated molecular patterns (PAMPs) that are expressed on infectious agents, and mediate the production of cytokines necessary for the development of effective immunity. The various TLRs exhibit different patterns of expression. This receptor is most abundantly expressed in placenta, and in myelomonocytic subpopulation of the leukocytes. It has been implicated in signal transduction events induced by lipopolysaccharide (LPS) found in most gram-negative bacteria. Mutations in this gene have been associated with differences in LPS responsiveness. Also, several transcript variants of this gene have been found, but the protein coding potential of most of them is uncertain. [provided by RefSeq

Sequence: DGKSWNPEGTVGTGCNWQEATSI

Specifications

Antigen TLR4
Applications Dot Blot, ELISA, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against synthetic peptide of TLR4.
Dilution ELISA (1:20000) Dot Blot (1:20000) Western Blot (1:500) The optimal working dilution should be determined by the end user.
Formulation In buffer containing 0.02% sodium azide
Gene TLR4
Gene Alias ARMD10/CD284/TOLL/hToll
Gene Symbols TLR4
Host Species Rabbit
Immunogen A synthetic peptide corresponding to amino acids 970-1000 of human TLR4.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7099
Target Species Human
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.