Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TRPV2 Rabbit anti-Human, Rat, Polyclonal Antibody, Abnova™

Catalog No. 89117000 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-117-000 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-117-000 Supplier Abnova Corporation Supplier No. PAB14319
Only null left
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against synthetic peptide of Trpv2.

Sequence: KNSASEEDHLPLQVLQSP

Specifications

Antigen TRPV2
Applications Immunofluorescence, Immunohistochemistry, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against synthetic peptide of Trpv2.
Dilution Western Blot (0.5-1 ug) The optimal working dilution should be determined by the end user.
Formulation Lyophilized from PBS
Gene Trpv2
Gene Alias MGC105451/Vrl1
Gene Symbols Trpv2
Host Species Rabbit
Immunogen A synthetic peptide (conjugated with KLH) corresponding to amino acids 744-761 of rat Trpv2.
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 29465
Test Specificity Immunohistochemical analysis in rat dorsal root ganglia and spinal cord indicates a high level of specificity for this antiserum.
Target Species Human, Rat
Content And Storage Store at 4°C on dry atmosphere. After reconstitution with 50μL of deionized water store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Form Lyophilized
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.