Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Rabbit anti-Turkey IgG (H+L) Secondary Antibody, Novus Biologicals™
SDP

Catalog No. NBP173813 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
2 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP173813 2 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP173813 Supplier Novus Biologicals Supplier No. NBP173813
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

IgG (H+L) Polyclonal specifically detects IgG (H+L) in Turkey samples. It is validated for ELISA, Immunohistochemistry, Western Blot.

Specifications

Antigen IgG (H+L)
Applications ELISA, Immunohistochemistry, Western Blot
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1:2000 - 1:10000, ELISA 1:20000 - 1:100000, Immunohistochemistry 1:1000 - 1:5000
Host Species Rabbit
Immunogen Turkey IgG whole molecule
Purification Method Delipidation and Defibrination
Quantity 2 mL
Regulatory Status RUO
Research Discipline Adaptive Immunity, Immunology
Primary or Secondary Secondary
Test Specificity This product was prepared from monospecific antiserum by a delipidation and defibrination. Assay by immunoelectrophoresis resulted in a single precipitin arc against Turkey IgG and Turkey Serum.
Reconstitution Reconstitute with 2.0 ml deionized water (or equivalent).
Target Species Turkey
Content And Storage Store lyophilized antibody at 4°C. Aliquot reconstituted liquid and store at -20°C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.