Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

UBA1 Rabbit anti-Dog, Human, Mouse, Rabbit, Rat, Polyclonal Antibody, Abnova™

Catalog No. 89114994 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
500 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-114-994 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-114-994 Supplier Abnova Corporation Supplier No. PAB11287
Only null left
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against full-length recombinant UBA1.

The protein encoded by this gene catalyzes the first step in ubiquitin conjugation to mark cellular proteins for degradation. This gene complements an X-linked mouse temperature-sensitive defect in DNA synthesis, and thus may function in DNA repair. It is part of a gene cluster on chromosome Xp11.23. Alternatively spliced transcript variants that encode the same protein have been described. [provided by RefSeq

Specifications

Antigen UBA1
Applications ELISA, Immunohistochemistry (PFA fixed), Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against full length recombinant UBA1.
Dilution ELISA (1:2000-1:10000) Western Blot (1:1000-1:5000) Immunohistochemistry (2-20 ug/mL) The optimal working dilution should be determined by the end user.
Formulation In 20mM KH2PO4, 150mM NaCl, pH 7.2 (0.01% sodium azide)
Gene UBA1
Gene Alias A1S9/A1S9T/A1ST/AMCX1/GXP1/MGC4781/SMAX2/UBA1A/UBE1/UBE1X
Gene Symbols UBA1
Host Species Rabbit
Immunogen Recombinant GST fusion protein corresponding to full length human UBA1.
Quantity 500 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7317
Target Species Canine, Human, Mouse, Rabbit, Rat
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.