Learn More
Immunology Consultants Affinity Puriied Rabbit anti-VSV-g Tag Antibody, 0.1 mg
SDP
Supplier:  Immunology Consultants RVV45AZ
Encompass_Preferred
Rabbits were immunized with highly purified YTDIEMNRLGK (VSV-g epitope) and the resulting antiserum was collected. Antibody was immunoaffinity purified off a peptide containing immunosorbent. Antibody concentration was determined using an absorbance at 280 nm: 1.4 equals 1.0 mg of IgG. This antibody reacts with YTDIEMNRLGK (VSV-g epitope) as determined by IEP and ELISA techniques. It is suitable for blotting, ELISA and IHC applications. Optimal working dilutions should be determined experimentally by the investigator.
Catalog No. 50-196-2142
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.