Learn More
Biomatik Corporation Rabbit Bone Morphogenetic Protein 2 (BMP2) CLIA Kit
Supplier:  Biomatik Corporation EKU09521
Encompass_Preferred
Rabbit Bone Morphogenetic Protein 2 (BMP2) CLIA Kit for detecting BMP2 in serum, plasma, tissue homogenates, other biological fluids. Species Reactivity: Rabbit (Oryctolagus cuniculus). Detection Range: 13.7-10,000pg/ml. Sensitivity: 6.4pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Chemiluminescence. Assay Time: 2h, 40min. Accession Number: O46564. Gene ID: 100009349. Target Synonyms: BMP2A; BMP-2A; Hemochromatosis Modifier. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-6821
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.