Learn More
ACROBiosystems Rabbit Complement C2 200ug
Supplier:  ACROBiosystems CO2R82E3200UG
Encompass_Preferred
Biotinylated Rabbit Complement C2 Protein HisAvitag (CO2-R82E3) is expressed from human 293 cells (HEK293). It contains AA Thr 21 - Leu 744 (Accession # G1SS66).
Catalog No. 50-253-5227
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.