Learn More
Biomatik Corporation Rabbit Complement Component 5a (C5a) ELISA Kit
Supplier:  Biomatik Corporation EKU03410
Encompass_Preferred
Rabbit Complement Component 5a (C5a) ELISA Kit for detecting C5a in serum, plasma, other biological fluids. Species Reactivity: Rabbit (Oryctolagus cuniculus). Detection Range: 6.25-400pg/ml. Sensitivity: 2.82pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: G1SPF9. Gene ID: 103352022. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-9038
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.