Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Rabbit IgG Isotype Control
Rabbit Polyclonal Isotype Control
Supplier: Invitrogen™ 10500C

Description
Rabbit IgG Isotype Control for Flow, Ctrl
Rabbit IgG refers to immunoglobulin G antibodies produced by rabbits in response to an antigen. These antibodies are commonly used in laboratory research as a detection tool for specific proteins, molecules, or cells. Rabbit IgG antibodies are highly specific and have a strong affinity for their target, making them ideal for use in various applications such as western blotting, immunohistochemistry, and flow cytometry.
Specifications
Rabbit IgG | |
Polyclonal | |
Unconjugated | |
Immunoglobulin G; ImmunoglobulinG; Not Applicable | |
Normal Rabbit IgG | |
5 mL | |
Isotype Control (Primary) | |
4°C | |
Liquid | |
Responsibly packaged |
Control, Flow Cytometry | |
3 mg/mL | |
PBS with 0.1% sodium azide | |
Rabbit | |
Purified | |
RUO | |
Not Applicable | |
Isotype Control | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction