Learn More
Description
Specifications
Specifications
| Concentration | 1 mg/mL |
| For Use With (Application) | ELISA, Immunohistochemistry, SDS-PAGE, Western Blot |
| Formulation | 0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2 |
| Name | Rabbit IgG, Fc Protein |
| Quantity | 1 mg |
| Storage Requirements | Store at 4 C short term. Aliquot and store (at -20 C long term. Avoid freeze-thaw cycles.) |
| Cross Reactivity | Rabbit |
| Product Line | Novus |
| Purity or Quality Grade | Multi-step |
| Protein | Rabbit IgG, Fc |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
