Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Rabbit IgG (H+L) Antibody, R&D Systems™
GREENER_CHOICESDP

Catalog No. D301CABS2 Shop All Novus Products
Change view
Click to view available options
Quantity:
0.5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D301CABS2 0.5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. D301CABS2 Supplier R&D Systems Supplier No. D301CABS2
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Donkey Polyclonal Antibody

IgG Polyclonal specifically detects IgG in Rabbit samples. It is validated for Secondary.

Specifications

Antigen IgG
Applications Not Available
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Hidden
Formulation Lyophilized from a 0.2 μm filtered solution in NaH2PO4, NaCl and Sodium Azide with Trehalose. with No Preservative
Gene Alias Immunoglobulin G, ImmunoglobulinG
Gene Symbols IGHG1
Host Species Donkey
Purification Method Affinity Purified
Quantity 0.5 mg
Regulatory Status RUO
Primary or Secondary Secondary
Gene ID (Entrez) 3500
Test Specificity This antibody reacts with heavy chains on rabbit IgG, and with light chains on all rabbit immunoglobulins. No reactivity observed with other serum proteins, or IgG from bovine, chicken, goat, guinea pig, horse, human, mouse, rat, sheep, or Syrian hamster, by IEP. This antibody adsorbed against solid phase human IgG, bovine IgG, mouse IgG, rat IgG, and goat IgG.
Reconstitution Sterile PBS to a final concentration of 0.2 mg/mL.
Target Species Rabbit
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 °C to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution.
Product Type Antibody
Isotype IgG
Product Line R&D Systems
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.