Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Rabbit IgG Isotype Control, Alexa Fluor™ 488, Novus Biologicals™
SDP

Rabbit Antibody

Supplier:  Novus Biologicals NBP236463AF488

Catalog No. NBP236463X


Only null left
Add to Cart

Description

Description

IgG specifically detects IgG in Rabbit samples. It is validated for ELISA, Immunohistochemistry.
Specifications

Specifications

IgG
Alexa Fluor 488
50mM Sodium Borate with 0.05% Sodium Azide
IGHG1
Multi-step
RUO
3500
Rabbit
Liquid
ELISA, Immunohistochemistry, SDS-Page
ELISA, Immunohistochemistry
Immunoglobulin G, ImmunoglobulinG
Rabbit
500 Tests
Isotype Control (Primary)
Rabbit IgG whole molecular was prepared from normal serum by a multi-step process which includes delipidation, salt fractionation and ion exchange chromatography followed by extensive dialysis against the buffer. Rabbit IgG whole molecular was assayed by immunoelectrophoresis resulted in a single precipitin arc against anti-Rabbit IgG and anti-Rabbit Serum.
Store at 4C in the dark.
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.

For Research Use Only