Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Rabbit IgG Isotype Control, FITC, eBioscience™

Catalog No. 501128945
Encompass_Preferred
Change view
Click to view available options
Quantity:
25 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-112-8945 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-112-8945 Supplier Invitrogen™ Supplier No. 11461480
Add to Cart
Edge
Add to Cart

Rabbit Polyclonal Isotype Control

Description: The rabbit IgG polyclonal antibody is useful as an isotype control immunoglobulin. Applications Reported: The Rabbit IgG isotype control has been reported for use as a negative control for immunofluorescent staining and flow cytometric analysis. Applications Tested: This isotype control antibody has been tested by flow cytometric analysis. Please use at the same concentration as the experimental antibody. Excitation: 488 nm; Emission: 520 nm; Laser: Blue Laser. Filtration: 0.2 μm post-manufacturing filtered.

Rabbit IgG refers to immunoglobulin G antibodies produced by rabbits in response to an antigen. These antibodies are commonly used in laboratory research as a detection tool for specific proteins, molecules, or cells. Rabbit IgG antibodies are highly specific and have a strong affinity for their target, making them ideal for use in various applications such as western blotting, immunohistochemistry, and flow cytometry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Rabbit IgG
Applications Control, Flow Cytometry
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate FITC
Formulation PBS with 0.09% sodium azide; pH 7.2
Gene Alias Immunoglobulin G; ImmunoglobulinG; Not Applicable
Host Species Rabbit
Purification Method Affinity chromatography
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Isotype Control (Primary)
Target Species Not Applicable
Content And Storage 4°C, store in dark, DO NOT FREEZE!
Product Type Isotype Control
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.