Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Rabbit IgG Isotype Control

Catalog No. 086199
Encompass_Preferred
Change view
Click to view available options
Quantity:
18 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
08-619-9 18 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 08-619-9 Supplier Invitrogen™ Supplier No. 086199
Add to Cart
Edge
Add to Cart

Rabbit Polyclonal Isotype Control

Rabbit IgG Isotype Control for IHC, Ctrl

Rabbit IgG refers to immunoglobulin G antibodies produced by rabbits in response to an antigen. These antibodies are commonly used in laboratory research as a detection tool for specific proteins, molecules, or cells. Rabbit IgG antibodies are highly specific and have a strong affinity for their target, making them ideal for use in various applications such as western blotting, immunohistochemistry, and flow cytometry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Rabbit IgG
Applications Control, Immunohistochemistry
Classification Polyclonal
Concentration 0.5 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA and 0.1% sodium azide; pH 7.4
Gene Alias Immunoglobulin G; ImmunoglobulinG; Not Applicable
Host Species Rabbit
Immunogen Rabbit Immunoglobins
Purification Method Purified
Quantity 18 mL
Regulatory Status RUO
Primary or Secondary Isotype Control (Primary)
Target Species Not Applicable
Content And Storage 4°C
Product Type Isotype Control
Form Liquid
Isotype Ig
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.