Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Rabbit IgG Recombinant Isotype Control (RbNP15), Alexa Fluor™ 488, eBioscience™

Catalog No. 53461682
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
53-461-682 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 53-461-682 Supplier Invitrogen™ Supplier No. 53461682
Add to Cart
Edge
Add to Cart

Rabbit Recombinant Monoclonal Isotype Control

Description: This product is a rabbit monoclonal IgG and can be used as a matched isotype control for recombinant rabbit monoclonal antibodies. The RbNP15 IgG was selected as an isotype control following screening for low background on a variety of mouse and human cell types. Applications Tested: This rabbit monoclonal isotype control has been tested for use as a negative control for flow cytometric analysis. Please use at the same concentration as the experimental antibody. Excitation: 488 nm; Emission: 519 nm; Laser: Blue Laser Filtration: 0.2 μm post-manufacturing filtered.

Rabbit IgG refers to immunoglobulin G antibodies produced by rabbits in response to an antigen. These antibodies are commonly used in laboratory research as a detection tool for specific proteins, molecules, or cells. Rabbit IgG antibodies are highly specific and have a strong affinity for their target, making them ideal for use in various applications such as western blotting, immunohistochemistry, and flow cytometry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Rabbit IgG
Applications Control, Flow Cytometry
Classification Recombinant Monoclonal
Clone RbNP15
Concentration 0.5 mg/mL
Conjugate Alexa Fluor 488
Formulation PBS, proprietary buffer with 0.09% sodium azide; pH 7.2
Gene Alias Immunoglobulin G; ImmunoglobulinG; Not Applicable
Host Species Rabbit
Purification Method Protein A
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Isotype Control (Primary)
Target Species Not Applicable
Content And Storage 4°C, store in dark, DO NOT FREEZE!
Product Type Isotype Control
Form Liquid
Isotype IgG
Product Line eBioscience
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.