Learn More
Invitrogen™ Rabbit IgG Recombinant Isotype Control (RbNP15), eBioscience™
Rabbit Recombinant Monoclonal Isotype Control
Supplier: Invitrogen™ 14461682

Description
Description: This product is a rabbit monoclonal IgG and can be used as a matched isotype control for recombinant rabbit monoclonal antibodies. The RbNP15 IgG was selected as an isotype control following screening for low background on a variety of mouse and human cell types. Applications Tested: This rabbit monoclonal isotype control has been tested for use as a negative control for flow cytometric analysis. Please use at the same concentration as the experimental antibody. Filtration: 0.2 μm post-manufacturing filtered.
Rabbit IgG refers to immunoglobulin G antibodies produced by rabbits in response to an antigen. These antibodies are commonly used in laboratory research as a detection tool for specific proteins, molecules, or cells. Rabbit IgG antibodies are highly specific and have a strong affinity for their target, making them ideal for use in various applications such as western blotting, immunohistochemistry, and flow cytometry.
Specifications
Rabbit IgG | |
Recombinant Monoclonal | |
0.5 mg/mL | |
PBS with 0.09% sodium azide; pH 7.4 | |
Rabbit | |
100 μg | |
Isotype Control (Primary) | |
4°C | |
Liquid | |
Responsibly packaged |
Control, Flow Cytometry | |
RbNP15 | |
Unconjugated | |
Immunoglobulin G; ImmunoglobulinG; Not Applicable | |
Protein A | |
RUO | |
Not Applicable | |
Isotype Control | |
IgG |
Safety and Handling
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.