Learn More
Enzo Life Sciences Goat anti-rabbit IgG, polyclonal antibody (AP conjugate) (1 ml)
Supplier: Enzo Life Sciences ADISAB301J
Host: Goat. Immunogen: Purified rabbit IgG (whole molecule). Species Reactivity: Rabbit. Specificity: Based on IEP and/or ELISA, reacts with whole molecule rabbit IgG, and also the light chains of other rabbit immunoglobulins. No antibody detected against non-immunoglobulin serum proteins. Antibody tested by ELISA and/or solid-phase adsorbed to ensure minimal cross-reaction with human serum proteins. Applications: ELISA, WB. Recommended dilutions: ELISA (1:2,500-25,000). Western Blot (1:2500-25,000, colorimetric). Suggested dilutions/conditions may not be available for all applications. Optimal conditions must be determined individually for each application. Purity: Immunoaffinity purified. Formulation: Liquid. In 0.005M TRIS-hydrogen chloride, pH 8.0, containing 0.125M sodium chloride, 7.5mg/ml BSA (IgG-free, protease-free), 50% glycerol, and 0.025M sodium azide.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.