Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Rabbit IgG VisUCyte HRP Polymer Antibody, R&D Systems™
GREENER_CHOICE

Goat Polyclonal Antibody

$245.00 - $2360.00

Specifications

Antigen IgG
Dilution Immunohistochemistry
Applications Immunohistochemistry
Classification Polyclonal
Conjugate HRP Polymer
View More Specs

Products 4
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
VC003025
GREENER_CHOICE
View Documents
R&D Systems
VC003025
25 mL
Each for $245.00
Only null left
Add to Cart
 
VC003050
GREENER_CHOICE
View Documents
R&D Systems
VC003050
50 mL (2 x 25 mL)
Each for $456.00
Only null left
Add to Cart
 
VC003125
GREENER_CHOICE
View Documents
R&D Systems
VC003125
125 mL
Each for $1,218.00
Only null left
Add to Cart
 
VC003250
GREENER_CHOICE
View Documents
R&D Systems
VC003250
250 mL (2 x 125 mL)
Each for $2,360.00
Only null left
Add to Cart
 
Description

Description

IgG Polyclonal specifically detects IgG in Rabbit samples. It is validated for Immunohistochemistry.
Specifications

Specifications

IgG
Immunohistochemistry
HRP Polymer
RUO
Supplied as a solution in PBS with preservative.
3500
IgG
Affinity Purified
Detects heavy ( gamma ) chains on rabbit IgG and light chains on all rabbit immunoglobulins, based on IEP. No cross-reactivity is observed with non-immunoglobulin rabbit serum proteins or serum proteins from human, mouse or rat, based on IEP. VisUCyteÖ HRP Polymer provides high sensitivity with low background in IHC.
Immunohistochemistry
Polyclonal
Goat
Rabbit
Immunoglobulin G, ImmunoglobulinG
Rabbit IgG
Secondary
6 months from date of receipt, 2 to 8 degreesC as supplied.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.