Learn More
Kingfisher Biotech Inc Rabbit IL-16 Recombinant Protein
SDP
Supplier:  Kingfisher Biotech Inc RP1433U005
Encompass
The Rabbit IL-16 yeast-derived recombinant protein is not tagged and is naturally endotoxin-free. The purity is greater than 98% as visualized by SDS-PAGE analysis. Rabbit IL-16 applications are for cell culture, ELISA standard, and Western Blot Control. Rabbit IL-16 yeast-derived recombinant protein can be purchased in multiple sizes. Rabbit IL-16 Specifications: (Molecular Weight: 12.7 kDa) (Amino Acid Sequence: GLSPSSDSAA APSAASDVPA ESATEAAICT VTLEKTSAGL GFSLEGGRGS LRGDKPITIN RIFKGVASGP SATLQPGDEI LHVAGTALQG LTRFEAWNII KALPDGPVTV VIRRGLQPQG TVATGDP (127)) (Gene ID: 100356333). For research use only. Made in the USA
Catalog No. 50-253-3264
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.