Learn More
Kingfisher Biotech Inc Rabbit LIF Recombinant Protein
SDP
Supplier:  Kingfisher Biotech Inc RP1011U005
Encompass
The Rabbit LIF yeast-derived recombinant protein is not tagged and is naturally endotoxin-free. The purity is greater than 98% as visualized by SDS-PAGE analysis. Rabbit LIF applications are for cell culture, ELISA standard, and Western Blot Control. Rabbit LIF yeast-derived recombinant protein can be purchased in multiple sizes. Rabbit LIF Specifications: (Molecular Weight: 19.7 kDa) (Amino Acid Sequence: SPLPINPVNA TCNTHHPCPS NLMSQIRSQL AQLNGTANAL FILYYTAQGE PFPNNLDKLC GPNVTDFPPF HANGTEKVRL VELYRIVAYL GTALGNITRD QKTLNPTAHS LHSKLNATAD TLRGLLSNVL CRLCSKYHVA HVDVAYGPDT SGKDVFQKKK LGCQLLGKYK QVMAVLAQAF (180)) (Gene ID: 100358914). For research use only. Made in the USA
Catalog No. 50-241-3452
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.