Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abcam Rabbit Multiclonal Antibody, Anti-NOTCH3, RM1102, Carrier Free, 100 μg

Catalog No. 502698314
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-269-8314 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-269-8314 Supplier Abcam Supplier No. AB315349100UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Rabbit multiclonal antibody for NOTCH3 staining in flow cytometry, IHC, and western blot, featuring a carrier-free, conjugation-ready formulation.

  • Host Species: Rabbit
  • Clonality: Multiclonal
  • Isotype: IgG
  • Applications: WB, IHC-P, Flow Cyt (Intra)
  • Reactivity: Human
  • Purification: Affinity purification Protein A
  • Form: Liquid
  • Buffer: PBS, pH 7.2 to 7.4
  • Features: Carrier free, conjugation ready

The product is subject to the following: Abcam Restricted Use Statement

Specifications

Target Molecule NOTCH3
Reactivity Human
Label or Dye Unconjugated
Detection Location In-Blot Detection, In-Cell Detection
Form Liquid
Buffer PBS, pH 7.2 to 7.4
Cross Reactivity Human
Formulation PBS, pH 7.2 to 7.4
Quantity 100 μg
For Use With (Application) Flow Cytometry, IHC-P, Western Blot
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.