Learn More
Kingfisher Biotech Inc Rabbit OPN Recombinant Protein
SDP
Supplier:  Kingfisher Biotech Inc RP2091U025
Encompass
The Rabbit Osteopontin yeast-derived recombinant protein is not tagged and is naturally endotoxin-free. The purity is greater than 98% as visualized by SDS-PAGE analysis. Rabbit Osteopontin applications are for cell culture, ELISA standard, and Western Blot Control. Rabbit Osteopontin yeast-derived recombinant protein can be purchased in multiple sizes. Rabbit Osteopontin Specifications: (Molecular Weight: 33.4 kDa) (Amino Acid Sequence: LPVKHADSGS SEEKQLYHKH PDALATWLNP DPSQKQNLLT PQNAMSSEEK DDLKQETLPS KSIESHDHMD DIDEDEDDDH VDNRDSNESD DADHPDDSHH SDESHQSDES DEVTVYPTED AATTVFTEVV PTVETYDGRG DSVAYRLKRS KSKMFHVSNA QYPGASEEDL SSHVDSEDLD DTPRAIPVAQ HLNVPSDWDS QEKDSHDVSQ VDDHSVETQS HEQARQYKRE ANDNSVEHSH SIDSQESSKV SQESQSREFR SHEDKLAIEP KSEEDEEHRQ LRVSHELDST SSEIN (295)) (Gene ID: 100008982). For research use only. Made in the USA
Catalog No. 50-253-2449
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.