Learn More
Innovative Research Inc Rabbit Red Blood Cells 10ml
Catalog No. 502034860
Encompass_Preferred
Rabbit Red Blood Cells from Innovative Research are manufactured using whole blood collections from Rabbits located in the USA. The whole blood is washed to remove the platelet-rich plasma, buffy coat layer, and leukocytes. Red blood cells are perishable and are collected and processed upon receipt of order. We offer reliable, consistent products that deliver reliable, consistent results.
Catalog No. 50-203-4860 Supplier Innovative Research Inc Supplier No. IRBRBC10ML
Rabbit Red Blood Cells from Innovative Research are manufactured using whole blood collections from Rabbits located in the USA. The whole blood is washed to remove the platelet-rich plasma, buffy coat layer, and leukocytes. Red blood cells are perishable and are collected and processed upon receipt of order. We offer reliable, consistent products that deliver reliable, consistent results.