Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Rabenosyn 5 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA560853
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA560853 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA560853 Supplier Invitrogen™ Supplier No. PA560853
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: DSPAPNPFSE EDEHPQQRLS SPLVPGNPFE EPTCINPFEI DSDSGPEAEE PIEEELLLQQ IDNIKAYIFD AKQC Highest antigen sequence identity to the following orthologs: Mouse - 76%, Rat - 80%.

This gene encodes a zinc finger transcription factor. The encoded protein likely plays a role in transcriptional repression of interleukin 2. Mutations in this gene have been associated with posterior polymorphous corneal dystrophy-3 and late-onset Fuchs endothelial corneal dystrophy. Alternatively spliced transcript variants encoding different isoforms have been described.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Rabenosyn 5
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Rbsn
Gene Accession No. Q9H1K0
Gene Alias 110 kDa protein; 5330426D11Rik; FLJ34993; FYVE finger-containing Rab5 effector protein rabenosyn-5; MGC126210; RAB effector RBSN; Rabenosyn 5; rabenosyn, RAB effector; rabenosyn-5; Rbsn; ZFYVE20; zinc finger FYVE domain-containing protein 20; zinc finger, FYVE domain containing 20
Gene Symbols Rbsn
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human Rabenosyn 5. Recombinant protein control fragment (Product #RP-96334).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 64145
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.