Learn More
TARGETMOL CHEMICALS INC RABEPRAZOLE SODIUM 100MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T1651
Encompass_Preferred
Also available in 5 mg 10 mg 25 mg 50 mg 200 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Rabeprazole sodium (Habeprazole Sodium) is a 4-(3-methoxypropoxy)-3-methylpyridinyl derivative of timoprazole that is used in the therapy of stomach ulcers and Zollinger-Ellison syndrome. The drug inhibits H(+)-K(+)-exchanging ATPase which is found in gastric parietal cells. purity: 99%
Catalog No. 50-258-1173
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.