Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Rabeprazole sodium salt, Thermo Scientific™

Catalog No. p-7227401
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Specifications

Chemical Name or Material Rabeprazole sodium salt
CAS 117976-90-6
Quantity 1 g
Solubility Soluble in chloroform, ethanol, ethylacetate, methanol; inSoluble in ether, hexane
Melting Point 145°C to 155°C
Shipping Condition No special requirements
Product Line Life science
Molecular Formula C18 H20 N3 Na O3 S
Physical Form Powder
Color White to Yellow to Beige
Purity ≥97.5%
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.