Learn More
TARGETMOL CHEMICALS INC RABEXIMOD 5MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T34249
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg and bulk. Please contact Fisher for quotes. Rabeximod is a potent immunomodulator that reduces the severity of autoimmune diseases in rat models. Rabeximod inhibits arthritis in a time-dependent manner by stimulating TLR2 and TLR4 downstream to block the activation of inflammatory cells most likely macrophages. Rabeximod effectively reduces brain antigen presentation in mice during anti-inflammatory therapy for traumatic brain injury. purity: 100%
Catalog No. 50-258-0274
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.