Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RABGGTB Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA528324
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA528324 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA528324 Supplier Invitrogen™ Supplier No. PA528324
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Recommended positive controls: U87-MG, SK-N-SH, IMR32, SK-N-AS, Mouse brain, Rat brain. Predicted reactivity: Mouse (96%), Rat (96%), Zebrafish (91%), Xenopus laevis (91%), Pig (98%), Sheep (93%), Rhesus Monkey (100%), Bovine (98%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

RABGGTB catalyzes the transfer of a geranyl-geranyl moiety from geranyl-geranyl pyrophosphate to both cysteines in Rab proteins with an -XXCC, -XCXC and -CCXX C-terminal, such as RAB1A, RAB3A and RAB5A respectively.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RABGGTB
Applications Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7
Gene RABGGTB
Gene Accession No. P53611, P53612, Q08603
Gene Alias fj12b07; geranylgeranyl transferase type II subunit beta; Geranylgeranyl transferase type-2 subunit beta; GGTase-II-beta; Ggtb; rab geranylgeranyl transferase b subunit; Rab geranylgeranyl transferase componenet, subunit beta; Rab geranylgeranyl transferase, b subunit; Rab geranylgeranyltransferase beta subunit; rab geranylgeranyltransferase subunit beta; rab geranyl-geranyltransferase subunit beta; Rab geranylgeranyltransferase, beta subunit; rab GG transferase beta; rab GGTase beta; Rabggtb; Type II protein geranyl-geranyltransferase subunit beta; wu:fj12b07; zgc:56443; zgc:56443 protein
Gene Symbols RABGGTB
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human RABGGTB. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 19352, 25533, 5876
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.