Learn More
Invitrogen™ Rabies Virus Matrix Protein Monoclonal Antibody (1F9H6)

Description
Rabies Virus Matrix Protein Monoclonal Antibody for Western Blot, ELISA
Specifications
Specifications
| Antigen | Rabies Virus Matrix Protein |
| Applications | ELISA, Western Blot |
| Classification | Monoclonal |
| Clone | 1F9H6 |
| Concentration | 1.19 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.03% Proclin 300; pH 7.4 |
| Gene Alias | Matrix protein; Phosphoprotein M2 |
| Host Species | Mouse |
| Immunogen | Recombinant Rabies virus Matrix protein (1-202AA). |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.