Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Rabies Virus Monoclonal Antibody (C94A)
GREENER_CHOICE

Catalog No. MA17079
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MA17079 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MA17079 Supplier Invitrogen™ Supplier No. MA17079
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

MA1-7079 detects the Rabies virus. This monoclonal antibody is broadly reactive with over 20 different strains including: Duwenhage, Mokola & Lagosbat. MA1-7079 has been successfully used in ELISA, immunofluorescence, and immunocytochemistry procedures. The MA1-7079 immunogen is Rabies virus lysate. This antibody is specific for a surface glycoprotein.

Rabies virus (Neurotropic lyssavirus) is a member of the Rhabdoviridae family. Rabies is a single stranded, neurotropic, negative sense RNA virus which encodes 5 proteins: a glycoprotein, a nucleoprotein, and three others. The mature virus has a bullet shape, a protein coat, and a lipid envelope. The outer surface of the virus is covered with thumblike glycoprotein projections 5-10 nm long and 3 nm in diameter. The virus averages approximately 780 nm in length. Virus infectivity is destroyed by lipid solvents. Rabies virus is a very successful virus, with a very wide range of hosts. It causes an acute, central nervous system infection, characterized by CNS irritation, followed by paralysis and death. Approximately 50,000 human deaths each year are caused by rabies.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Rabies Virus
Applications ELISA, Immunocytochemistry
Classification Monoclonal
Clone C94A
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% sodium azide; pH 7.2
Gene Alias Neurotropic lyssavirus
Host Species Mouse
Immunogen Rabies virus lysate.
Purification Method Protein A
Quantity 1 mL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Virus
Content And Storage 4°C
Product Type Antibody
Form Liquid
Isotype IgG2a
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.