Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Abcam rac-alpha-Tocopherol, Vitamin E Form, Liquid, 1 mL
Description
- Molecular Weight: 430.7 Da
- Purity: >95%
- Form: Liquid
- Function: Fat-soluble antioxidant and peroxyl radical scavenger that protects polyunsaturated fatty acids in membranes and lipoproteins
- Anticarcinogenic properties
- Storage Buffer: Constituents: 92.5% Hexane, 7.5% (2R)-2,5,7,8-tetramethyl-2-[(4R,8R)-4,8,12-trimethyltridecyl]-3,4-dihydrochromen-6-ol
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Specifications
| Form | Liquid |
| Product Type | Antioxidant |
| Content And Storage | Store at -20°C. |
| Buffer | 92.5% hexane, 7.5% (2R)-2,5,7,8-tetramethyl-2-[(4R,8R)-4,8,12-trimethyltridecyl]-3,4-dihydrochromen-6-ol |
| Target Molecule | Polyunsaturated Fatty Acids |
| Shipping Condition | Ambient |
| Formulation | 92.5% Hexane, 7.5% (2R)-2,5,7,8-tetramethyl-2-[(4R,8R)-4,8,12-trimethyltridecyl]-3,4-dihydrochromen-6-ol |
| Quantity | 1 mL |
| For Use With (Application) | Antioxidant research, membrane protection, lipoprotein protection, anticarcinogen research |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?