Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ rac-CCT 250863, >98%
GREENER_CHOICESDP

Catalog No. 45465
Change view
Click to view available options
Quantity:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
45-465 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 45-465 Supplier Tocris Bioscience™ Supplier No. 4546/5

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Potent Nek2 inhibitor

Potent Nek2 inhibitor (IC50 = 73 nM). Exhibits selectivity for Nek2 over PLK1, MPS1, Cdk2 and Aurora A.

Specifications

CAS 1364269-06-6
Quantity 5 mg
Solubility Soluble to 100mM in 1eq. HCl and to 100mM in DMSO
Molecular Formula C24H25F3N4O2S
Formula Weight 490.54
Purity >98%
Molecular Weight (g/mol) 490.545
Chemical Name or Material 4-[2-Amino-5-[4-[(dimethylamino)methyl]-2-thienyl]-3-pyridinyl]-2-[[(2Z)-4,4,4-trifluoro-1-methyl-2-
Recommended Storage Store at -20°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.