Learn More
TARGETMOL CHEMICALS INC RAC-WAY-161503 10MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T13332
Encompass_Preferred
Also available in 5 mg 25 mg 50 mg 100 mg and bulk. Please contact Fisher for quotes. (Rac)-WAY-161503 is a selective and high-affinity agonist of the 5-HT2C receptor (Ki 4 nM; EC50 12 nM) with anti-obesity and antidepressant effects. purity: 98%
Catalog No. 50-257-7397
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.