Description
Includes
- Anti-Rac1, clone 23A8
- Mg2+ Lysis/Wash Buffer, 5X
- 100X GTPγS, 10mM
- 100X GDP, 100mM
- Rac/cdc42 Assay Reagent (PAK1 PBD, agarose)
Specifications
Specifications
| For Use With (Application) | Affinity Binding Assay |
| Quantity | 30 Assays/Kit |
| Type | Activation Assay Kit |