Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RAC1 Monoclonal Antibody (1F4)
GREENER_CHOICE

Catalog No. PIMA532928
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA532928 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA532928 Supplier Invitrogen™ Supplier No. MA532928
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

RAC1 Monoclonal Antibody for Western Blot, ICC/IF, IHC (P), Flow

The protein encoded by this gene is a GTPase which belongs to the RAS superfamily of small GTP-binding proteins. Members of this superfamily appear to regulate a diverse array of cellular events, including the control of cell growth, cytoskeletal reorganization, and the activation of protein kinases. Two transcript variants encoding different isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RAC1
Applications Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Monoclonal
Clone 1F4
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% sodium azide; pH 7.2
Gene Rac1
Gene Accession No. P63000, P63001, Q6RUV5
Gene Alias AL023026; cell migration-inducing gene 5 protein; D5Ertd559e; GTPase; MGC111543; MIG5; p21-Rac1; Rac 1b; Rac family small GTPase 1; RAC1; Rac-1; Rac2; rac2 GTP-binding protein; ras-like protein TC25; RAS-related C3 botulinum substrate 1; ras-related C3 botulinum toxin substrate 1; ras-related C3 botulinum toxin substrate 1 (rho family small GTP binding protein Rac1); ras-related C3 botulinum toxin substrate 1 (rho family, small GTP binding protein Rac1); rho family, small GTP binding protein Rac1; TC25; TC-25
Gene Symbols Rac1
Host Species Mouse
Immunogen This antibody is generated from a mouse immunized with a KLH conjugated synthetic peptide between amino acids from human.
Purification Method Protein G
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 19353, 363875, 5879
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.