Learn More
Description
The protein encoded by this gene is a GTPase which belongs to the RAS superfamily of small GTP-binding proteins. Members of this superfamily appear to regulate a diverse array of cellular events, including the control of cell growth, cytoskeletal reorganization, and the activation of protein kinases. Two transcript variants encoding different isoforms have been found for this gene.
- Theoretical MW (kDa): 46.86
- Preparation Method: In vitro wheat germ expression system
- Purification: Glutathione Sepharose 4 fast flow
- Storage Buffer: 50 mM Tris-HCI, 10 mM reduced Glutathione, pH=8.0 in the elution buffer
Sequence: MQAIKCVVVGDGAVGKTCLLISYTTNAFPGEYIPTVFDNYSANVMVDGKPVNLGLWDTAGQEDYDRLRPLSYPQTDVFLICFSLVSPASFENVRAKWYPEVRHHCPNTPIILVGTKLDLRDDKDTIEKLKEKKLIPITYPQGLAMAKEIGAVKYLECSALTQRGLKTVFDEAIRAVLCPPPVKKRKRKCLLL
Best when used within three months from the date of receipt.
ELISA, Western Blotting (Recombinant Protein), Antibody Production, Protein Array
Specifications
Specifications
| Accession Number | AAH04247 |
| For Use With (Application) | Antibody Production, Protein Array, ELISA, Western Blot |
| Formulation | 50mM Tris HCl, 10mM reduced Glutathione, pH 8 in the Elution Buffer |
| Gene ID (Entrez) | 5879 |
| Molecular Weight (g/mol) | 46.86 |
| Name | RAC1 (Human) Recombinant Protein (P01) |
| pH Range | 8 |
| Preparation Method | In vitro wheat germ expression system |
| Purification Method | Glutathione Sepharose 4 Fast Flow |
| Quality Control Testing | 12.5% SDS-PAGE Stained with Coomassie Blue. |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.