Learn More
Abcam Rac2 Activation Assay Kit, Human, Mouse, 20 Tests
Description
- Reactive species: Mouse, Human
- Sample types: Tissue, Suspension cells, Adherent cells
- Application: Measure RAC2 in cell and tissue extracts
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Specifications
| Target Molecule | Rac2 |
| No. of Reactions | 20 Tests |
| Format | Kit |
| Product Type | Western Blot Kit |
| Detection Location | In-Solution Detection |
| Detection Method | Colorimetric |
| Cross Reactivity | Mouse, Human |
| Sample Type | Tissue, Suspension Cells, Adherent Cells |
| Quantity | 20 Tests |
| For Use With (Application) | RAC2 measurement |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.