Learn More
Brooks Automation Rack Adaptor
Catalog No. PSSRTA173 Shop All Brooks Automation Products
Change view
Click to view available options
Includes:
10 x 6, Alinity CI
2 x Haematology Racks
Abbott Architect 7 Way
Abbott Carrier Tray
Hitachi/Stago Star Evo
Inpeco Rack
Suit 4 x Sysmex
Tecan Evo 16 Way
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Includes
PSSRTA173 10 x 6, Alinity CI
PSSRTA026 Suit 4 x Sysmex
PSSRTA047 Abbott Architect 7 Way
PSSRTA050 Inpeco Rack
PSSRTA055 Hitachi/Stago Star Evo
PSSRTA059 Abbott Carrier Tray
PSSRTA063 Tecan Evo 16 Way
PSSRTA138 2 x Haematology Racks
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Catalog No. PSSRTA173 Supplier Brooks Automation Supplier No. PSSRTA173
Add to Cart
Add to Cart
Accessory that allows the PathFinder system to interface with different destination analyzer racks
  • Helps sample tubes which are sorted, capped or archived, transfers into racks and fit your laboratory analyzers or storage system
  • The adapter bridges the standard PathFinder rack format to the analyzer-specific rack format
  • Adapter enables the PathFinder to load/unload racks that match the analyser or storage system your lab uses
Specifications
Type Rack Adapter
Includes 10 x 6, Alinity CI