Learn More
Thermo Scientific RACK CARTRIDGE 6ML
Supplier:  Thermo Scientific 79211
Encompass_Preferred
General Accessories for Centrifuges; Cartridge rack; Thermo Scientific; Accommodates 2mL and 6mL Ultracrimp cartridges as well as 5.1mL Quick-Seal cartridges; Each
Catalog No. 50-867-245