Learn More
Brooks Automation Rack Fence Assembly, Specimen Tray
Catalog No. PSSRTA072 Shop All Brooks Automation Products
Change view
Click to view available options
Includes:
Stat Fitted
Catalog No. Includes
PSSRTA072 Stat Fitted
1 options
Catalog No. PSSRTA072 Supplier Brooks Automation Supplier No. PSSRTA072
Add to Cart
Add to Cart
Sample tube racks designed for automated handling for laboratory use
  • Use on PathFinder where sample tube racks are placed or moved
  • Fixes the position of the rack/tray on the deck of the instrument, ensuring correct orientation and preventing mis-placement of tubes during automation
  • An accessory component to the specimen tray that ensures proper alignment, ejecting or receiving racks, perhaps for safety or positioning purposes
  • Sub-assembly used in the tray module of the automation system
Specifications
Type Rack Fence Assembly
Includes Stat Fitted