Learn More
Julabo USA Inc Each test tube rack, 60 tubes, 15mm diameter, polypropylene
SDP
Supplier:  Julabo USA Inc 8970380
Encompass_Preferred
Bath Accessories, Julabo, Test tube racks, for 60 test tubes, 16/17 mm, diameter, to +80°C. For use with shaking water bath models SW22, SW23. Requires basic tray 8970369. Polypropylene.
Catalog No. 50-753-4065
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.