Learn More
Julabo USA Inc RACK FOR 90 UL TUBES 11/12MM
SDP
Supplier:  Julabo USA Inc 8970382
Encompass_Preferred
Polyproylene test tube rack for 90 tubes, 11/12 mm diameter, to +80 �C. For use with JULABO shaking water baths SW22, SW23. Requires test tube rack carrier tray: 8970369.
Catalog No. 50-753-4067
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.