Learn More
Thermo Scientific RACK FREEZER CRYOPLUS 23-7/8
Supplier:  Thermo Scientific 4000002
Encompass_Preferred
Rack, Special Purpose; Thermo Scientific; Cryo/Cryoplus inventory racks; Stainless steel; Holds box no. 11 containers; W x D x H: 5.5 x 5.5 x 23.875 in.
Catalog No. 50-874-27
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.