Learn More
Research Products International Corp Floating Bubble Rack with Pressure Plate, Capacity 8 1.5ml Tubes, Fits 400ml Beaker
SDP
Supplier:  Research Products International Corp 143008
Encompass_Preferred
Specially designed racks feature a polypropylene plate that prevents micro-tube lids from opening during boiling or high temperature incubation. Racks float to maintain sample tubes immersed at a constant depth. Supplied one each.
Catalog No. 50-212-264
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.