Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ZMYND8 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA554282
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA554282 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA554282 Supplier Invitrogen™ Supplier No. PA554282
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: TTKTDKTSTT GSILNLNLDR SKAEMDLKEL SESVQQQSTP VPLISPKRQI RSRFQLNLDK TIESCKAQLG INEISEDVYT AVEHSDSEDS EKSDSSDSEY ISDDEQKSKN Highest antigen sequence identity to the following orthologs: Mouse - 92%, Rat - 92%.

The protein encoded by this gene is a receptor for activated C-kinase (RACK) protein. The encoded protein has been shown to bind in vitro to activated protein kinase C beta I. In addition, this protein is a cutaneous T-cell lymphoma-associated antigen. Finally, the protein contains a bromodomain and two zinc fingers, and is thought to be a transcriptional regulator. Multiple transcript variants encoding several different isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZMYND8
Applications Immunohistochemistry (Paraffin), Immunocytochemistry, Western Blot
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ZMYND8
Gene Accession No. Q9ULU4
Gene Alias 1110013E22Rik; 2010005I16Rik; 3632413B07Rik; AI316811; AL024039; CTCL tumor antigen se14-3; CTCL-associated antigen se14-3; Cutaneous T-cell lymphoma-associated antigen se14-3; KIAA1125; predicted protein of HQ2893; Prkcbp1; PRO2893; protein kinase C binding protein 1; protein kinase C-binding protein 1; Rack7; spikar; spikar delta C; zinc finger MYND domain-containing protein 8; zinc finger MYND-type containing 8; zinc finger, MYND-type containing 8; Zmynd8
Gene Symbols ZMYND8
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human RACK7. Recombinant protein control fragment (Product #a href=.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23613
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.