Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

United Scientific™ Racked Micro Tips, Sterile
SDPSCI_ED

Catalog No. S8538028 Shop All United Scientific Supplies, Inc Products
Change view
Click to view available options
Volume (Metric):
10 μL
200 μL
1000 μL
Color:
Clear Blue
Natural
Yellow
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric) Color
S8538028 1000 μL Clear Blue
S8538025 10 μL Natural
S8538027 200 μL Natural
S8538026 200 μL Yellow
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. S8538028 Supplier United Scientific™ Supplier No. UNP10113
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

These Sterile Racked Micro Tips are disposable tips intended for use with most brands of pipettors.

  • The highly smooth, hydrophobic surface provides low liquid retention and increased accuracy and repeatability.
  • Inner radial rings enable an airtight grip on the tip cone.
  • Manufactured in a precision molding facility from 100% virgin polypropylene.
  • Free of DNase, RNase and pyrogen.

Specifications

Description Racked Micro Tips, Sterile, 100 to 1000 μL, Blue
Volume (Metric) 1000 μL
Filtered Non-filtered
Sterility Sterile
Color Clear Blue
Rack Color Blue
Autoclavable Autoclavable
Model UNP10113
Certifications/Compliance USP Class VI certified
For Use With (Application) Pipetting
Sterilization Method Steam
Format Rack, 10 x 96
Tip Style Standard
Packaging 10 Racks of 96
Quantity 960 Pk.
Product Type Micro Tips
Disposable Reusable
For Use With (Equipment) Universal Fit
Graduated No
Material Polypropylene
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.