Learn More
Thermo Scientific™ Racks and Inserts for Refrigerated and Heated Bath Circulators
These stainless-steel racks and inserts support a range of tubes and are designed for use with Thermo Scientific™ Refrigerated and Heated Bath Circulators.
Supplier:  Thermo Scientific™ 1600003
 View more versions of this product
Catalog No. 13-255-80
Only null left
Add to Cart
Description
Description
These stainless-steel racks and inserts support a range of tubes and are designed for use with Thermo Scientific™ Refrigerated and Heated Bath Circulators.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
A5B,A10B A24B,S49,S19T,S14P,S21P
Rack Insert
100-10 mm Tubes
Stainless Steel
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.