Learn More
Thermo Scientific™ Racks for the CryoMed™ Controlled-Rate Freezer
Description
More flexibility for your controlled-rate freezing applications with Thermo Scientific™ Racks for the CryoMed™ Controlled-Rate Freezer. Includes an assortment of cane, canister, straw and vial racks.
- Warranty varies by country, please contact us for details
Specifications
Specifications
| Color | Silver |
| Description | Five 1.2 mL/2 mL Cryo Racks (Small) |
| For Use With (Equipment) | All CryoMed controlled-rate freezers |
| Holds | 5 x 1.2/2 mL Cryo Racks (Small) |
| Type | Vial Rack Holder |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.